Learn More
Fisherbrand™ MultiTube Vortexers: Replacement Tray Pad Set
Shop All Fisherbrand Products
Click to view available options
For Use With (Equipment):
Multitube vortexer
Unit Size:
Each
Description
Replacement parts for Fisherbrand™ MultiTube Vortexers.
For use with Fisher Scientific MultiTube Vortexers
- Replacement tray pad set includes top and lower pads
- Gray
Specifications
Specifications
| Product Type | Tray Pad Set |
| Color | Gray |
| For Use With (Equipment) | Multitube vortexer |