Learn More
Fisherbrand™ Baume Hydrometers
Shop All Fisherbrand Products
Click to view available options
Certifications/Compliance:
Without 3-point certification
Increments:
0.1°
Length (Metric):
290 mm
295 mm
305 mm
310 mm
Baume Range:
19 to 31°
29 to 41°
40 to 51°
49 to 61°
60 to 70°
9 to 21°
Unit Size:
Each
Description
Baume heavy scale for working with liquids heavier than water.
- Permanently marked with individual serial numbers
Order Info
Use Fisherbrand cylinder 08-530J with 250mL of liquid for submersion, available separately
Certifications
A certificate is provided to indicate traceability to standards provided by NIST™.
Compliance
Accuracy meets or exceeds ASTM™ E 100 specifications, Calibrated according to ASTM E 126 specifications.
Specifications
Specifications
| Type | Baume Hydrometer |
| Increments | 0.1° |
| Certifications/Compliance | Without 3-point certification |
| Length (Metric) | 295 mm |
| Material | Soda Lime Glass |
| Baume Range | 60 to 70° |
| For Use With (Application) | Baume 'Heavy' Scale For Liquids Heavier than Water |