Learn More
Fisherbrand™ Adjustable Jacobs Chuck
Accepts stirrer shafts from 1/16 in. to 3/8 in. (1.5 mm to 10mm) in diameter.
Brand:  Fisherbrand™ 37020-9036
Product Code. 15192997
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Fits 3/8 in. (10mm) diameter motor shafts
  • Accepts stirrer shafts from 1/16 in. to 3/8 in. (1.5 mm to 10mm) in diameter
Specifications
Specifications
10mm dia. Motor Shaft
Adjustable Jacobs Chuck