Description
- Fits 3/8 in. (10mm) diameter motor shafts
- Accepts stirrer shafts from 1/16 in. to 3/8 in. (1.5 mm to 10mm) in diameter
Specifications
10mm dia. Motor Shaft | |
Adjustable Jacobs Chuck |