missing translation for 'onlineSavingsMsg'
Learn More

Ekastu Safety™ Mandil Filtering Facepiece FFP3/V with Cool Down Exhalation Valve

Protects against toxic and highly toxic particles up to 50 times the NPF value. EKASTU Safety™ Mandil Filtering Facepiece FFP3/V with Cool Down Exhalation Valve has a five-year shelf life in original packaging.

Brand:  Ekastu Safety™ 412183

Product Code. 10574162

  • € 38.00 / Pack of 5
Estimated Shipment: 25-06-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • With Cool Down exhalation valve
  • With colour-coded nosepiece and exhalation valve
  • Pprotection against toxic and highly toxic particles up to 50 times the NPF value
  • Continous adjustable harness
  • Broad cotton-lined inner sealing lip guarantees close and comfortable fit as well as safety
  • Filter class DIN EN 149:2001 FFP3 NR
  • Specially designed for wearer of EKASTU spectacles and goggles
  • In original packaging: 5 years shelf life
  • 5 pieces per pack
Specifications

Specifications

Red
160 l/min, constant flow max. 3.0 mbar
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ekastu Safety™ Mandil Filtering Facepiece FFP3/V with Cool Down Exhalation Valve >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.