Learn More
Epredia™ Fine Pore Cassettes
Product Code. 16374086 Shop All Epredia Products
Click to view available options
Color:
Beige
Blue
Gray
Green
Lavender
Light Green
Lilac
Orange
Pink
Tan
White
Yellow
Unit Size:
Pack of 1000
Product Code. 16374086
Brand: Epredia™ B851735BL
For very small biopsy tissue samples, cassettes have a front-edge lid hinge with attached lids that are available in 12 different color varieties
Printing
  • Surface finish and reinforced print faces are engineered to optimize print quality
  • Tube design is precisely engineered to reduce jamming and prevent downtime
Grossing, Processing and Embedding
  • Cassette design and flow characteristics tested to ensure efficient reagent exchange
  • Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples
Sectioning
  • Precision engineering for solid connection to microtomes
Archiving
  • Cassette surface texture optimizes scannability for archiving
TRUSTED_SUSTAINABILITY
Specifications
Pore Style Fine Pore
Closure Included Yes
Packaging Type Loose Packed
Quantity 1000/Cs.
Closure Type Attached Lid, Front Hinge
PrintMate Compatible No
Color Blue
Product Type Cassette