Learn More
Cole-Parmer™ Lab Markers
Shop All Cole-Parmer Products
Click to view available options
Color:
Blue
Red
Unit Size:
Pack of 10
Description
- Fine-point fiber tip
- Quick-drying ink
- Writes on cold, wet surfaces
- Available in four different colors
Specifications
Specifications
| Color | Red |
| For Use With (Application) | For Glass, Porcelain or Metal surfaces |
| Quantity | 10/Pk. |