Learn More
Shamrock™ Film Welder w/ Knife Seam, 300mm
Seals and cleanly cuts in one easy step. Shamrock™ Film Welder with Knife Seam 300mm is an easy-to-use, all-in-one unit - simply place bag between seal bars, press down and release; trim excess with the equipped slide cutter.
Brand:  Shamrock™ 44839
Encompass
Product Code. 10789182
  • POA / Each
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Explore more special offers
Description
Description
  • Seam length of 300mm
  • Moulding time of 0.2/1.3 seconds
  • One-hand operation
  • Compact design
Specifications
Specifications
Film Welder w/ Knife
Seals and cleanly cuts in one easy step
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Shamrock™ Film Welder w/ Knife Seam, 300mm