Learn More
Thermo Scientific™ Fiberboard Box Dividers for Ultra-Low Temperature and Cryogenic Freezers
Organize vial storage within cryo boxes with these fiberboard box dividers.
POA - € 33.47
Product Code | Brand | Holds | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Holds | Quantity | Price | Quantity & Availability | ||||
12744388
|
Thermo Scientific™
5959 |
49 (7 x 7 Grid) | 12 Pack |
€ 13.87
Each |
Estimated Shipment: 03-04-2025
Log in to see stock. |
||||
15810530
|
Thermo Scientific™
5960 |
64 (8 x 8 Grid) | 12 Pack |
€ 32.86
Pack of 12 |
In Stock
Log in to see stock. |
||||
11849490
|
Thermo Scientific™
AVL6212 |
81 (9 x 9 Grid) | 12 Pack |
€ 33.47
Pack of 12 |
In Stock
Log in to see stock. |
||||
10059500
|
Thermo Scientific™
6212 |
81 (9 x 9 Grid) | 12 Pack | N/A | |||||
15907976
|
Thermo Scientific™
820025 |
25 (5 x 5 Grid) | Each |
€ 4.52
Each |
In Stock
Log in to see stock. |
||||
15821152
|
Thermo Scientific™
820049 |
49 (7 x 7 Grid) | Each |
€ 4.52
Each |
In Stock
Log in to see stock. |
||||
11329341
|
Thermo Scientific™
820081 |
81 (9 x 9 Grid) | Each |
€ 4.62
Each |
In Stock
Log in to see stock. |
||||
Description
Organize vial storage with Thermo Scientific™ Fiberboard Box Dividers for Ultra-Low Temperature and Cryogenic Freezers.
- Use with fiberboard storage boxes to divide box into grids for vial storage
- Box dividers for cold storage application available in 7 x 7 , 8 x 8, 9 x 9 or 10 x 10 grids