Learn More
Fiacitabine, MedChemExpress
MedChemExpress Fiacitabine(NSC 382097; FIAC; FOAC) is a selective inhibitior of DNA replication of herpes simplex virus(HSV) with IC50 values of 2.5 nM and 12.6 nM for HSV1 and HSV2, respectively.
Brand: Medchem Express HY-50735
This item is not returnable.
View return policy
Description
Fiacitabine(NSC 382097Specifications
Fiacitabine | |
Off-White | |
C9H11FIN3O4 | |
NSC 382097 FIAC FOAC | |
O=C(N=C1N)N([C@@H]([C@H]([C@@H]2O)F)O[C@@H]2CO)C=C1I | |
371.1 | |
Research | |
COVID-19-anti-virus |
69123-90-6 | |
Research | |
10 mg | |
DMSO : ≥ 37 mg/mL (99.70 mM) | |
371.1 | |
98.83% | |
Solid |
Research purposes only