Learn More
Gibco™ Fetal Bovine Serum, Qualified One Shot™, Australia origin
Click to view available options
Quantity:
10 x 50 mL
Unit Size:
Pack of 10
Description
If you’re one of the more than 80% of researchers who aliquots their FBS into 50mL conical tubes, your life is about to change for the better. Transferring your FBS takes away your control — of how you spend your time, your contamination risk, and your volume consistency. It also wastes costly material.
The new Gibco™ One Shot™ FBS 50mL bottle design eliminates the need to aliquot, and the variability that comes with it. And that improves your chances for consistent results.
Product Features:
The new Gibco™ One Shot™ FBS 50mL bottle design eliminates the need to aliquot, and the variability that comes with it. And that improves your chances for consistent results.
Product Features:
- Wide mouth, chamfered edge, and curved shape for easy pouring and pipetting
- Engineered to freeze/thaw without splitting or breaking
- Fits in test tube racks and frees up freezer space
- No labeling mishaps with our easy-to-read package label has all the information you need
- Generates 33% less waste than aliquoting from 500mL bottles
Specifications
Specifications
| Country of Origin | Australia |
| Format | Bottle |
| Quantity | 10 x 50 mL |
| For Use With (Application) | Tissue Culture |