missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bohlender™ BOLA™ FEP Tubing
Perfect for aggressive and pure liquids or gases. FEP tubing features almost universal chemical resistance, and is free from extractable agents.
Brand: Bohlender™ S1815-20
This item is not returnable.
View return policy
Description
- Transparent, gastight tubing
- Non-porous
- Almost universal chemical resistance
- Free from extractable agents
- Physiologically safe
- Non-adhesive surface
- Very good sliding characteristics
- Very good dielectric characteristics
- Flame retardant according to UL94V0
- Oxygen value more than 95
- Resistant to irradiation and weather
- Can be sterilized in autoclaves
- Mechanical strength even at high temperatures
Specifications
FDA, USP Class VI | |
4 mm | |
6 mm | |
Perfect tubing for aggressive and pure liquids or gases | |
1 Pack | |
56 bar |
Autoclavable | |
FEP | |
1 mm | |
-270°C | |
Transparent | |
260°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction