Learn More
Fehling's reagent II for sugars, For determination of sugar, Reag. Ph. Eur., solution II: potassium sodium tartrate, alcaline, Honeywell Fluka™
Product Code. 15655040 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
250 mL
500 mL
1 L
1.5 L
2.5 L
6 x 250 mL
6 x 500 mL
30 L
4 x 2.5 L
6 x 1 L
Unit Size:
1 L
1.5 L
2.5 L
250mL
30 L
500mL
Pack of 4
Pack of 6
Product Code. 15655040
Brand: Honeywell-Fluka 36019250ML
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
for determination of sugar, Reag. Ph. Eur., solution II: potassium sodium tartrate, alcaline
  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Fehling’s reagent II for sugars
CAS 1310-73-2, 304-59-6, 7732-18-5
MDL Number MFCD00012467
UN Number UN1824
Density 1.23-1.25g/mL at 20°C
Quantity 250 mL