Learn More
Falcon™ Multiwell Library Storage Plate
GREENER_CHOICE
Ideal for storing, archiving, and transporting of pharmaceutical drug candidates
Brand:  Falcon™ 351190
324.77 EUR valid until 2025-04-13
Use promo code "24122" to get your promotional price.
Product Code. 10693652
  • € 355.53 / Pack of 100
Estimated Shipment: 09-05-2025
to see stock.
Explore more special offers
Description
Falcon™ 96-well Polypropylene Microplate, not treated, round bottom, 25/pack, 50/case : The Falcon 96 well Polypropylene Library Storage Plate is convenient for compound storage : Notch H12
  • Exceedingly rigid and flat plate for consistent automated handling
  • Plate withstands repeated freeze-thaws down to -20°C
  • Low retention polypropylene and round-well design give low residual volumes after pipetting (<0.1 μL)
  • Raised well edges and flatness for heat or adhesive sealing
  • In addition to the round-bottom Falcon 96 well Library Storage Plate, V-bottom formats are also available.
  • TRUSTED_SUSTAINABILITY
    Specifications
    Specifications
    Clear
    Non-sterile
    100/Cs.
    Storage Microplate
    Polypropylene
    96
    25/Pk.
    Round
    Certificates
    Special Offers
    Special Offers
    Product Content Correction
    Product Title
    Falcon™ Multiwell Library Storage Plate > Round