missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mettler Toledo™ F2 OIML Class Advanced Weight (F2 PL C E)
Single F2 OIML weight includes a calibration certificate and is housed in a plastic box.
€ 119.00
Specifications
Product Type | Test Weight |
---|---|
Material | Stainless Steel |
Case Material | Plastic |
Density | 7950 (± 140) kg/m3 |
Includes | With certificate |
Description
- With calibration certificate
- Traceability Certificate: Delivered with a Calibration Certificate in accordance with ISO/IEC 17025 requirements
- Weight Calibration Performed by Robots: State-of-the-art robot technology minimizes errors and increases accuracy
- Weight with knob design have an adjusting cavity, which simplifies weight adjustment without the need for specialized equipment
Specifications
Test Weight | |
Plastic | |
With certificate |
Stainless Steel | |
7950 (± 140) kg/m3 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title