Learn More
Mettler Toledo™ F2 OIML Class Advanced Weight (F2 PL C E)
Single F2 OIML weight includes a calibration certificate and is housed in a plastic box.
Brand:  Mettler Toledo™ 30406424
 View more versions of this product
Product Code. 17710008
  • € 119.00 / Each
Estimated Shipment: 29-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • With calibration certificate
  • Traceability Certificate: Delivered with a Calibration Certificate in accordance with ISO/IEC 17025 requirements
  • Weight Calibration Performed by Robots: State-of-the-art robot technology minimizes errors and increases accuracy
  • Weight with knob design have an adjusting cavity, which simplifies weight adjustment without the need for specialized equipment
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Test Weight
Plastic
7950 (± 140) kg/m3
With certificate
Stainless Steel
1 g
Knob with Adjusting Cavity
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers