Learn More
behr Labor-Technik™ EZ 100 Soxhlet Extractor
Extract a desired compound with limited solubility in a solvent. behr Labor-Technik™ EZ 100 Soxhlet Extractor recycles a small amount of solvent to dissolve a larger amount of material.
Brand:  behr Labor-Technik™ B00217967
Product Code. 10340351
  • € 151.00 / Each
Estimated Shipment: 17-06-2025
to see stock.
Explore more special offers
Description
  • For 100mL extractions
Specifications
Specifications
Soxhlet Extractor
100 mL
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
behr Labor-Technik™ EZ 100 Soxhlet Extractor > Capacity: 100mL