Learn More
Bartels Rieger™ Barikos KS™ Eyewash Bottle
Intended for immediate flushing of eyes with water for eye injuries. Bartels Rieger™ Barikos KS™ Eyewash Bottles are filled with washing liquid in a sealed condition.
Brand: Bartels Rieger™ 954004
Description
Unlike conventional eye wash bottles, in which washing liquid is renewed frequently, the Barikos KS is filled and maintenance free. The water fillings consist of toxicologically harmless materials and conform to EN 15154-4.
- Red spray head and tilting valve
- Filled with 620 ml washing liquid, in a sealed condition 2 year shelf life, according to EN 15154-4
- CE mark according to the Medical Devices Set
- Ph-neutral eyewash waterborne without chemical additives
- Ergonomically shaped eye cavity
- Sterile and hygienic eyewash
- Universal and ready to use
- Lightweight and easy to use for flushing the eye in the first aid case with eye injuries
- Patented tilting valve with tubing allows upright and downwardly directed spray head
- Protective cap covers spray head; leak proof
Specifications
EN 15154-5, CE | |
620 mL Washing Liquid in Sealed Condition |