Learn More
Lenz Laborglasintrumente Extractors Acc. to Knöfler-Böhm
Product Code. 10214011
Click to view available options
Capacity (Metric):
30 mL
150 mL
250 mL
500 mL
Standard Taper Joint Bottom:
NS 29/32
NS 45/40
NS 60/46
Unit Size:
Each
Product Code. 10214011
Brand: Lenz Laborglasintrumente 05375049
Only null left
Add to basket
Add to basket
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • The complete apparatuses are consisting of: Jacketed extractor head made of DURAN™ tubing, Dimroth condenser (details on page 71)
  • With thread GL 14
  • Made of DURAN™ tubing
  • Round bottom flask
Specifications
Capacity (Metric) 250 mL
Material Duran Glass
Standard Taper Joint Bottom NS 45/40