missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell™ Extra™ 87-955 Series Orange Natural Rubber Latex Immersion Gloves
Provides for heavy duty handling where sensitivity is also required. Ansell Extra™ 87-955 Series Orange Natural Rubber Latex Immersion Gloves offer extremely high resistance to many ketones, salts, detergents, alcohols, alkalies and fats.
Brand: Ansell™ 87-955_9.5-10
This item is not returnable.
View return policy
Description
- 50% thicker than standard flocklined gloves
- Chlorinated for better grip and increased chemical resistance
- 100% natural rubber with no fillers for excellent resistance to mechanical hazards
- 100% cotton flocking for softness, comfort and increased absorption
- Ideal for chemical processing and preparation and maintenance of plant and heavy equipment
Specifications
Rubber Latex | |
Natural Rubber Latex | |
Immersion Gloves | |
87-955 | |
Orange | |
Cotton Flocking | |
Many ketones, salts, detergents, alcohols, alkalis and fats |
Natural Rubber Latex | |
No | |
10 | |
0.75 mm | |
Straight | |
144Pack | |
87-955 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction