missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ Extra™ 87-955 Series Orange Natural Rubber Latex Immersion Gloves

Provides for heavy duty handling where sensitivity is also required. Ansell Extra™ 87-955 Series Orange Natural Rubber Latex Immersion Gloves offer extremely high resistance to many ketones, salts, detergents, alcohols, alkalies and fats.

Brand:  Ansell™ 87-955_7.5-8

 View more versions of this product

Product Code. 15212879

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • 50% thicker than standard flocklined gloves
  • Chlorinated for better grip and increased chemical resistance
  • 100% natural rubber with no fillers for excellent resistance to mechanical hazards
  • 100% cotton flocking for softness, comfort and increased absorption
  • Ideal for chemical processing and preparation and maintenance of plant and heavy equipment
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Rubber Latex
Natural Rubber Latex
Immersion Gloves
87-955
Orange
Cotton Flocking
Many ketones, salts, detergents, alcohols, alkalis and fats
Natural Rubber Latex
No
8
0.75 mm
Straight
144Pack
87-955
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.