Learn More
Ansell™ AlphaTec™ 87-950 (ex EXTRA)
Product Code. 15272869 Shop All Ansell Products
Click to view available options
Size:
7.5
8
9
10
11
Unit Size:
12 pairs
144 pairs
Product Code. 15272869
Brand: Ansell™ 87950_8.59
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Extra protection for heavy duty applications.
  • 50% thicker than standard flocklined gloves.
  • Extremely high resistance to many ketones, salts, detergents, alcohols, alkalies and fats.
  • 100% natural rubber with no fillers. Excellent resistance to mechanical hazards plus increased chemical protection.
  • Chlorinated. For better grip and increased chemical resistance.
  • 100% cotton flocklining. Makes a heavy glove softer and more comfortable to wear. Helps absorb perspiration.
TRUSTED_SUSTAINABILITY
Specifications
Back Material Rubber Latex
Certifications/Compliance EN388: 2003, EN374:2003, EN388:2016, EN ISO 374-1:2016, EN ISO 374-5:2016
Cuff Material Natural Rubber Latex
Coating Material Natural Rubber Latex
Color Black
Latex Free No
Material Rubber Latex
Product Type Immersion Gloves
Size 9
Style 87-950
Thickness (Metric) 0.75 mm
For Use With (Application) For chemical processing, preparation and maintenance of plant and heavy equipment
Glove Cuff Style Straight
Liner Material Cotton Flocking
Quantity 12 Pairs
Resistant To Many ketones, salts, detergents, alcohols, alkalis and fats
Series 87-950
Length (Metric) 320 mm