missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ AlphaTec™ 87-950 (ex EXTRA)

Product Code. 15262869 Shop All Ansell Products
Click to view available options
Size:
7.5
8
9
10
11
Unit Size:
12 pairs
144 pairs
This item is not returnable. View return policy

Product Code. 15262869

Brand: Ansell™ 87950_7.58

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Extra protection for heavy duty applications.

  • 50% thicker than standard flocklined gloves.
  • Extremely high resistance to many ketones, salts, detergents, alcohols, alkalies and fats.
  • 100% natural rubber with no fillers. Excellent resistance to mechanical hazards plus increased chemical protection.
  • Chlorinated. For better grip and increased chemical resistance.
  • 100% cotton flocklining. Makes a heavy glove softer and more comfortable to wear. Helps absorb perspiration.
TRUSTED_SUSTAINABILITY

Specifications

Back Material Rubber Latex
Certifications/Compliance EN388: 2003, EN374:2003, EN388:2016, EN ISO 374-1:2016, EN ISO 374-5:2016
Cuff Material Natural Rubber Latex
Coating Material Natural Rubber Latex
Color Black
Latex Free Yes
Material Rubber Latex
Product Type Immersion Gloves
Size 8
Style 87-950
Thickness (Metric) 0.75 mm
For Use With (Application) For chemical processing, preparation and maintenance of plant and heavy equipment
Glove Cuff Style Straight
Liner Material Cotton Flocking
Quantity 12 Pairs
Resistant To Many ketones, salts, detergents, alcohols, alkalis and fats
Series 87-950
Length (Metric) 320 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.