Learn More
Ansell™ AlphaTec™ 87-950 (ex EXTRA)
Shop All Ansell Products
Click to view available options
Size:
7.5
8
9
10
11
Unit Size:
12 pairs
144 pairs
Description
- 50% thicker than standard flocklined gloves.
- Extremely high resistance to many ketones, salts, detergents, alcohols, alkalies and fats.
- 100% natural rubber with no fillers. Excellent resistance to mechanical hazards plus increased chemical protection.
- Chlorinated. For better grip and increased chemical resistance.
- 100% cotton flocklining. Makes a heavy glove softer and more comfortable to wear. Helps absorb perspiration.
Specifications
Specifications
| Back Material | Rubber Latex |
| Certifications/Compliance | EN388: 2003, EN374:2003, EN388:2016, EN ISO 374-1:2016, EN ISO 374-5:2016 |
| Cuff Material | Natural Rubber Latex |
| Coating Material | Natural Rubber Latex |
| Latex Free | No |
| Product Type | Immersion Gloves |
| Size | 11 |
| Style | 87-950 |
| Thickness (Metric) | 0.75 mm |
| Glove Cuff Color | Black |
| Show More |