Learn More
Memmert™ External Measuring Instrument
Product Code. 17199754 Shop All Memmert Products
Click to view available options
For Use With (Equipment):
HPP1060eco
HPP110eco
HPP1400eco
HPP2200eco
HPP260eco
HPP410eco
HPP750eco
ICO105
ICO150
ICO240
ICO50
IPP1060eco
IPP110eco
IPP1400eco
IPP2200eco
IPP260eco
IPP410eco
IPP750eco
Unit Size:
Each
Product Code. 17199754
Brand: Memmert™ HPP260ecoB04714
Only null left
Add to basket
Add to basket
Additional measuring head for temperature and humidity measurement
  • Additional measuring head
  • For temperature and humidity measurement
Specifications
For Use With (Equipment) HPP260eco
For Use With (Application) For Temperature and Humidity Measurement