Learn More
Buerkle™ Extension Rod
Shop All Bürkle Products
Click to view available options
Length (Metric):
500 mm
600 mm
1000 mm
Color:
Gray
Pure White
Silver
For Use With (Equipment):
ChemoSampler
Mole Drill Sampler
PTFE Scoop
PharmaPicker
PharmaPicker Pharma Sampler
Unit Size:
Each
Description
- Seven different drill bits are available, each of which is designed for a particular soil type
- For various applications
Specifications
Specifications
| Product Type | Extension Rod |
| Material | Stainless Steel |
| Autoclavable | Autoclavable |
| Description | Equipment for mole drill sampler |
| Color | Silver |
| For Use With (Equipment) | Mole Drill Sampler |
| Length (Metric) | 1000 mm |