Learn More
Novus Biologicals™ Exosome Standards (NCI-H1975 cell line)
Highly pure, lyophilized exosome standards with superior stability, optimal for multiple applications
Brand: Novus Biologicals™ NBP3-11688-60ug
Specifications
Store at 4°C. After reconstitution store at -70°C. | |
LYOPH | |
2 x 30 μg Vials |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Product Title
Novus Biologicals™ Exosome Standards (NCI-H1975 cell line) > 2 x 30ug Vials