Learn More
Novus Biologicals™ Exosome Standards (Human Plasma)
Highly pure, lyophilized exosome standards with superior stability, optimal for multiple applications
Brand: Novus Biologicals™ NBP3-11686-600ug
Specifications
Store at 4°C. After reconstitution store at -70°C. | |
LYOPH | |
6 x 100 μg Vials |
Product Content Correction
Product Title
Novus Biologicals™ Exosome Standards (Human Plasma) > 6 x 100ug Vials