Learn More
Novus Biologicals™ Exosome Standards (B16F10 cell line)
Product Code. 18650621 Shop All Bio Techne Products
Click to view available options
Quantity:
2 x 100 μg Vials
2 x 30 μg Vials
6 x 100 μg Vials
6 x 30 μg Vials
Unit Size:
Pack of 2
Pack of 6
Product Code. 18650621
Brand: Novus Biologicals™ NBP249866180ug
Highly pure, lyophilized exosome standards with superior stability, optimal for multiple applications
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store at 4°C. After reconstitution store at -70°C.
Concentration LYOPH
Quantity 6 x 30 μg Vials