Learn More
SI Analytics™ Exchange Unit
Shop All SI Analytics Products
Click to view available options
Volume (Metric):
5 mL
20 mL
Unit Size:
Each
Description
- Includes integrated chip for reagent data
- Comes with a brown glass bottle for titrant
- GL 45
- S 40 bottle adapter
- Includes tubes, drip tubes, and titration tip
Specifications
Specifications
| Color | Brown glass bottle for titrant |
| Container Size | GL 45 |
| Type | Exchange Unit |
| Includes | Integrated chip for reagent data |
| Volume (Metric) | 20 mL |