Learn More
EVOS™ EVOS™ Vessel Holder, KOVA™ Glasstic™ slide 10
Click to view available options
Quantity:
1 Holder
Unit Size:
Each
Description
- Fits microscope slide, cell culture flask or dish, or micro-well plate to stage of EVOS imaging systems for increased precision in sample alignment
- Accommodates one KOVA Glasstic Slide 10
- Easy-to-change vessel holders and stage plates available for this instrument platform
Specifications
Specifications
| Type | Vessel Holder |
| Product Line | EVOS |
| For Use With (Equipment) | EVOS™ XL Core Imaging System, EVOS™ FL Color Imaging System, EVOS™ XL Imaging System, EVOS™ FL Imaging System, EVOS™ FL Auto Imaging System |
| Format | 10 Slides |
| Holds | 1 KOVA™ Glasstic™ Slide 10 |
| Quantity | 1 Holder |