Learn More
A&D Weighing EK-i Series Compact Balance
EK-i Series Compact Balances feature a compact rugged design, reliable SG weighing technology, and versatile single and triple weighing ranges.
Brand:  A&D Weighing EK-2000i
343.71 EUR valid until 2024-12-31
Use promo code "21658" to get your promotional price.
 View more versions of this product
Product Code. 16631012
  • € 372.00 / Each
Explore more special offers
Description
Description
  • EC Type approved (EK-610i-EC, EK-6100i-EC)
  • Standard RS-232C interface
  • Large LCD display with backlight
  • Compact design
  • GLP compliant
  • Overload protection
  • Standard comparator function
  • Optional rechargeable battery pack
  • 9 Weighing units plus count and percentage mode
  • Counting function with Automatic Counting Accuracy Improvement (ACAI)
  • Full digital calibration with optional user definable mass setting
Specifications
Specifications
GLP/GMP/GCP/ISO
2000 g
0.1 g
EK-2000i Compact
RS-232 C
Backlit LCD
ABS, Stainless Steel
110/240 V to adapter (9 V, 670 mA), Ni-MH battery Pk. (option)
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers