missing translation for 'onlineSavingsMsg'
Learn More
Learn More
XPG Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-47577-25ul
This item is not returnable.
View return policy
Description
XPG Polyclonal specifically detects XPG in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| XPG | |
| Polyclonal | |
| Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| COFS3, DNA excision repair protein ERCC-5, DNA repair protein complementing XP-G cells, EC 3.1, ERCM2, excision repair cross-complementing rodent repair deficiency, complementationgroup 5, excision repair protein, xeroderma pigmentosum complementation group G protein, Xeroderma pigmentosum group G-complementing protein, xeroderma pigmentosum, complementation group G, XPGC, XPG-complementing protein, XPGUVDR | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| ERCC5 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: DDFSQYQLKGLLKKNYLNQHIEHVQKEMNQQHSGHIRRQYEDEGGFLKEVESRRVVSEDTSHYILIKGIQAKTVAEVDSESLPSSSKMHGMSFDVKSSPCE | |
| 25 μL | |
| Cancer, DNA Repair, Nucleotide Excision Repair | |
| 2073 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction