Learn More
Liebisch™ Evaporator with Fixed Heating Block for 60 Samples
€ 5800.00 - € 7070.00
Product Code | Brand | Temperature Range (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Temperature Range (Metric) | Price | Quantity & Availability | ||||||
11940188
|
Liebisch™
67386101 |
130°C |
€ 7070.00
Each |
Estimated Shipment: 09-05-2025
Log in to see stock. |
||||||
10198231
|
Liebisch™
67386201 |
200°C |
€ 5800.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Specifications
450 x 230 x 130 |
Product Title
Liebisch™ Evaporator with Fixed Heating Block for 60 Samples