Learn More
Lenz Laborglasintrumente Evaporating Flasks with Conical Ground Joint
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand: Lenz Laborglasintrumente 03042949
This item is not returnable.
View return policy
Description
- Especially for rotary evaporators
- With center neck and conical joint
- Pear-shaped
- With inscription label
- Material: DURAN™
Specifications
Duran Glass | |
NS 29/32 | |
250 mL |