Learn More
MedChemExpress Ethynylcytidine
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Specifications
Specifications
| Molecular Weight (g/mol) | 267.24 g/mol |
| Product Type | Reference compound |
| Content And Storage | Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month |
| Shipping Condition | Room temperature |
| Purity | 98.96% |
| Quantity | 5 mg |
| Solubility Information | DMSO : 250 mg/mL (935.49 mM∣Need ultrasonic) |
| For Use With (Application) | Cancer-programmed cell death |
| CAS | 180300-43-0 |
| Synonym | ECyD∣ TAS-106∣ 3'-C-Ethynylcytidine |