missing translation for 'onlineSavingsMsg'
Learn More

Ethylaluminium sesquichloride, 0.4M solution in hexane, AcroSeal™

Product Code. p-3212705 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
100mL
800mL
This item is not returnable. View return policy

Product Code. 10220793

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12075-68-2 | C6H15Al2Cl3 | 247.51 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12075-68-2, 110-54-3
Molecular Formula C6H15Al2Cl3
Molecular Weight (g/mol) 247.51
MDL Number MFCD00044852
InChI Key LDXMUMPEDOQULK-UHFFFAOYSA-K
Synonym ethylaluminum sesquichloride, ethylaluminium sesquichloride, sesquiethylaluminum chloride, easc, aluminum, trichlorotriethyldi, ethylaluminum sesquichioride, ethylaluminum sesquichloride solution, di-, i-chlorochlorotriethyldi-aluminum, chloro diethyl alumane; dichloro ethyl alumane
PubChem CID 25508
IUPAC Name chloro(diethyl)alumane;dichloro(ethyl)alumane
SMILES CC[Al](CC)Cl.CC[Al](Cl)Cl

Specifications

Color Colorless
Density 0.7010g/mL
Flash Point -22°C
Packaging AcroSeal™ Glass Bottle
Linear Formula (C2H5)2AlCl·Cl2AlC2H5
Quantity 100 mL
Specific Gravity 0.701
Solubility Information Solubility in water: vigorous reaction. Other solubilities: miscible with saturated aliphatic and aromatic,hydrocarbons
Formula Weight 247.51
Percent Purity 14.5 to 15.5% (on Al basis)
Physical Form Liquid
Chemical Name or Material Ethylaluminium sesquichloride
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Ethylaluminium sesquichloride, 0.4M solution in hexane
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
  • Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H361f-Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
  • H336-May cause drowsiness or dizziness.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P331-Do NOT induce vomiting.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.