missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethanolamine benzoate hydrochloride, 98+%, Thermo Scientific Chemicals
€ 116.00 - € 329.00
Chemical Identifiers
CAS | 545375-30-2 |
---|---|
Molecular Formula | C9H12ClNO2 |
Molecular Weight (g/mol) | 201.65 |
MDL Number | MFCD09953460 |
InChI Key | ZXTMLOSVFWKYCL-UHFFFAOYSA-N |
Synonym | ethanolamine benzoate hydrochloride, 2-aminoethyl benzoate hydrochloride, ethanolamine benzoate hydrochloride, 98+%, 2-benzoyloxyethylamine hydrochloride |
PubChem CID | 67778770 |
IUPAC Name | 2-aminoethyl benzoate;hydrochloride |
SMILES | Cl.NCCOC(=O)C1=CC=CC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11475600
|
Thermo Scientific Alfa Aesar
H25864.14 |
25 g |
€ 116.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11485600
|
Thermo Scientific Alfa Aesar
H25864.22 |
100 g | N/A | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
545375-30-2 | |
201.65 | |
ZXTMLOSVFWKYCL-UHFFFAOYSA-N | |
67778770 | |
Cl.NCCOC(=O)C1=CC=CC=C1 |
C9H12ClNO2 | |
MFCD09953460 | |
ethanolamine benzoate hydrochloride, 2-aminoethyl benzoate hydrochloride, ethanolamine benzoate hydrochloride, 98+%, 2-benzoyloxyethylamine hydrochloride | |
2-aminoethyl benzoate;hydrochloride |
Specifications
545375-30-2 | |
C9H12ClNO2 | |
ethanolamine benzoate hydrochloride, 2-aminoethyl benzoate hydrochloride, ethanolamine benzoate hydrochloride, 98+%, 2-benzoyloxyethylamine hydrochloride | |
Cl.NCCOC(=O)C1=CC=CC=C1 | |
201.65 | |
201.65 | |
Ethanolamine benzoate hydrochloride |
141°C to 142°C | |
MFCD09953460 | |
ZXTMLOSVFWKYCL-UHFFFAOYSA-N | |
2-aminoethyl benzoate;hydrochloride | |
67778770 | |
≥98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only