Learn More
Ethanol 70% in Water Solution For Laboratory Use, Chem Lab
Product Code. 16320638
Click to view available options
Ethanol 70%, 5 L
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.
5L Ethanol 70% in Water solution 700ml C2H5OH + 1% C3H8O + 1% C4H8O + DBN + H2O to 1L For laboratory use
5LT Ethanol 70% in Water solution 700ml C2H5OH + 1% C3H8O + 1% C4H8O + DBN + H2O to 1L For laboratory use
Specifications
Melting Point -117°C
CAS 64-17-5
Color Colorless
Density 0.85 g/mL
Boiling Point 78°C
Quantity 5 L
Flash Point 12 °C
Packaging Polyethylene
Physical Form Liquid
Molecular Formula C2H6O
Water 0.28
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
SMILES CCO
IUPAC Name ethanol
Molecular Weight (g/mol) 46.07
Show More Show Less
compliance-icons
Product Identifier
  • Ethanol 70% in Water solution
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
  • MIXTURE LIST-Contains : ethanol