Learn More
Notrax ESD Wrist Band
missing translation for 'orderingAttributeHoverText'
Wrist Band; For Use With: Connect a Worker's Skin to the Ground
Description
- Wrist Bands connect a worker’s skin to ground and are therefore the primary means of controlling static charge on personnel
- When properly worn and connected to ground, static charges are safely dissipated from the person
- Meets the requirements of EOS/ESD S1, MIL-STD-1686, and EIA 625
- Specialized line of ESD accessories to ensure a static free workplace and protect employees and sensitive electrical equipment
- Use these accessories to connect table work surface, employees and conductive material to a common grounding point
- This creates a balance between all items and personnel to ensure static electricity will flow to ground in a controlled manner
- For use in ESD working zones to protect electrostatic sensitive components
Specifications
Specifications
Product Type | Wrist Band |
Material | Wire |
For Use With (Application) | Connect a Worker's Skin to the Ground |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Product Title
Notrax ESD Wrist Band >