Learn More
Eppendorf™ S-4-104 Rotor Adapters
Available in a wide range of configurations to suit a variety of applications.
Brand: Eppendorf™ 5825741005
319.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Includes: 2 piece set
Description
- For use with Centrifuge 5810/5810 R
Specifications
Rotor Adapter | |
1 x 250 mL Tubes | |
S-4-104 rotor for 5810/5810R centrifuge | |
2 Pack |
Product Content Correction
Product Title
Eppendorf™ S-4-104 Rotor Adapters > For 1 x 250mL tubes