Learn More
Memmert™ Entry Port Right (Individual Location)
Product Code. 17120235 Shop All Memmert Products
Click to view available options
For Use With (Equipment):
HPP1060eco
HPP110eco
HPP1400eco
HPP2200eco
HPP260eco
HPP410eco
HPP750eco
IPP1060eco
IPP110eco
IPP1400eco
IPP2200eco
IPP260eco
IPP410eco
IPP750eco
Unit Size:
Each
Product Code. 17120235
Brand: Memmert™ IPP410ecoF5
Only null left
Add to basket
Add to basket
Entry port with flap closure and custom TBC position in right side
  • Not in combination with illumination units
  • Moisture tight
  • Can be closed by flap and silicone stopper
Specifications
Color Clear
For Use With (Equipment) IPP410eco
Diameter (Metric) 23 mm