Learn More
Memmert™ Entry Port Left Centre/Top
Product Code. 17150105 Shop All Memmert Products
Click to view available options
For Use With (Equipment):
HPP1060eco
HPP110eco
HPP260eco
HPP410eco
HPP750eco
IPP1060eco
IPP110eco
IPP260eco
IPP410eco
IPP750eco
Unit Size:
Each
Product Code. 17150105
Brand: Memmert™ IPP110ecoF1
Only null left
Add to basket
Add to basket
For introducing connections at the side and can be closed by flap
  • Standard position left centre/top
  • Not for modelsize 110 with light module
Specifications
Color Clear
For Use With (Equipment) IPP110eco
Diameter (Metric) 23 mm