Learn More
Memmert™ Entry Port Custom Position
Product Code. 17110115 Shop All Memmert Products
Click to view available options
For Use With (Equipment):
HPP1060eco
HPP110eco
HPP260eco
HPP410eco
HPP750eco
IPP1060eco
IPP110eco
IPP260eco
IPP410eco
IPP750eco
Unit Size:
Each
Product Code. 17110115
Brand: Memmert™ IPP110ecoF7
Only null left
Add to basket
Add to basket
Silicone Entry Port flap and stopper closure custom position TBC in the back
  • Individual position at the back (please, state location; not in combination with illumination units)
  • Moisture tight, can be closed by silicone stopper
Specifications
Color Clear
For Use With (Equipment) IPP110eco
Material Silicone
Diameter (Metric) 40 mm