Learn More
Memmert™ Entry Port 23 mm, Standard Position Side Right Side Centre/Top, For Hcp150
Product Code. 16334827 Shop All Memmert Products
Click to view available options
Diameter (Metric):
23 mm
Unit Size:
Each
Product Code. 16334827
Brand: Memmert™ HCP150F3
Only null left
Add to basket
Add to basket
Use these entry ports with convection ovens. Memmert™ Convection and Universal Oven Entry Points can be closed by flap in special positions at the back.
  • Entry port with clear diameter
  • For Memmert Ovens
Specifications
For Use With (Equipment) Models 30, 55, 75, 110, 160, 260, 450, 750, 1060
Material Stainless Steel
Type Entry Port
Diameter (Metric) 23 mm