Description
Ensure quality and fitness for purpose with dedicated Thermo Scientific™ consumables, including Enhanced Matrix Tolerance (EMT) Center Tubes for Thermo Scientific™ iCAP™ Series ICP-OES systems.
Maintain optimum performance with quality consumablesAvailable EMT center tubes include:
- Quartz Center Tube, 1.0 mm
- Quartz Center Tube, 1.5 mm
- Quartz Center Tube, 2.0 mm
- Ceramic Center Tube, 2.0 mm
All EMT center tubes are compatible with Thermo Scientific™ iCAP™ 7000 Series and iCAP™ PRO ICP-OES systems.
Product Title
Thermo Scientific™ Enhanced Matrix Tolerance (EMT) Center Tubes for the iCAP™ Series ICP-OES System