Learn More
Thermo Scientific™ Enhanced Matrix Tolerance (EMT) Center Tubes for the iCAP™ Series ICP-OES System
Maintain optimum performance of your Thermo Scientific™ iCAP™ Series ICP-OES system with Enhanced Matrix Tolerance (EMT) Center Tubes.
€ 249.00
Products 1
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
17893781
View Documents
Thermo Scientific™
BRE0019895
Quartz Center Tube, 2.0 mm
€ 249.00
Each
Description
Description
Ensure quality and fitness for purpose with dedicated Thermo Scientific™ consumables, including Enhanced Matrix Tolerance (EMT) Center Tubes for Thermo Scientific™ iCAP™ Series ICP-OES systems.
Maintain optimum performance with quality consumables
Available EMT center tubes include:
  • Quartz Center Tube, 1.0 mm
  • Quartz Center Tube, 1.5 mm
  • Quartz Center Tube, 2.0 mm
  • Ceramic Center Tube, 2.0 mm
All EMT center tubes are compatible with Thermo Scientific™ iCAP™ 7000 Series and iCAP™ PRO ICP-OES systems.
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Thermo Scientific™ Enhanced Matrix Tolerance (EMT) Center Tubes for the iCAP™ Series ICP-OES System