Learn More
Brady™ Engraved Replacement Labels for M610 and M611
Click to view available options
Color:
Black
Red
Silver
White
Yellow
Dimensions (LxW):
35 x 18 mm
45 x 15 mm
48 x 19 mm
Unit Size:
Each
Pack of 300
Description
- Engraved Plate Replacement labels for the M610 Portable Printer
- Aggressive adhesive and solvent resistance makes them a great replacement for engraved tags
- Service temperature of -40°C to +100°C
Specifications
Specifications
| Adhesive Type | Permanent Acrylic, Foam-backed |
| Coating | None |
| Color | Yellow |
| Finish | Gloss |
| Format | Label |
| Material | Polyester |
| Printable Sides | Single-side Printable |
| Print Technology | Thermal Transfer |
| Product Type | Engraved Plate Labels |
| Sterility | Non-sterile |
| Show More |