Learn More
Merck PureProteome™ Nickel Magnetic Beads
Click to view available options
Quantity:
10 mL
2 x 1 mL
Unit Size:
Each
Pack of 2
Description
- No lysate clearing—crude cell lysates are processed by magnetic beads
- Fast processing time—entire protocol is completed in less than one hour
- High capacity—bind 5 to 15mg of protein per mL of settled beads
- High affinity—binding can take place in buffer containing <5mM EDTA
- Flexible scale—varying the bead quantity allows different protein yields
- Flexible elution—native or denatured proteins
- Automatable—hands-free screening process
Specifications
Specifications
| Binding Property | 1 to 5.5mg/mL |
| Content And Storage | 2°C to 8°C |
| Particle Size | 10 μm |
| Quantity | 10 mL |
| For Use With (Application) | Purification of His-tagged recombinant Protein |
| Type | Magnetic Affinity Bead |
Product Content Correction
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur