Learn More
Merck 96-well V-bottom Collection Plate
Shop All Merck Products
Click to view available options
No. per Pack:
100
Unit Size:
Pack of 100
Specifications
Specifications
| Format | Standard |
| Array | 8 x 12 |
| Color | Clear |
| Material | Polypropylene |
| No. of Wells | 96 |
| Sterility | Non-sterile |
| Well Shape | Conical Bottom (V-well) |
| No. per Pack | 100 |
| Quantity | 100/Pk. |