Learn More
Motic™ ELWD Condenser for AE31E
Extra-long working distance condenser suitable for phase contrast. Motic™ ELWD Condenser for AE31E, when used with phase accessories, facilitates visualization of transparent specimens.
Brand: Motic™ 1101000202481
This item is not returnable.
View return policy
Description
- Concentrates and focuses light passing through the specimen.
- Extra-long working-distance condenser.
- Suitable for phase contrast.
Specifications
NA: 0.30 | |
ELWD Condenser | |
AE31E Microscopes | |
72 mm |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
Motic™ ELWD Condenser for AE31E > WD: 72mm