Learn More
Bel-Art™ Ellipso-Spoon and Spatula Samplers
Shop All SP Scienceware Products
Click to view available options
Capacity (Metric):
10 mL Bowl
70 mL bowl
Length (Metric):
15 cm
21 cm
30 cm
40 cm
50 cm
Size:
15 x 35 mm bowl
28 x 65 mm bowl
Unit Size:
Each
Description
- Crystals, powders, granulated and solid substances, and thick liquids retrieved
- Includes spoon end and spatula end
- Made of polished 304 grade stainless steel
Specifications
Specifications
| Autoclavable | Autoclavable |
| Capacity (Metric) | 10 mL Bowl |
| Product Type | Spoon and Spatula Sampler |
| Finish | Polished |
| For Use With (Equipment) | Bottles and containers |
| For Use With (Application) | Designed to extracts crystals, granulated and solid substances, thick liquids and delicate materials |
| Size | 15 x 35 mm bowl |
| Material | Stainless Steel |
| Length (Metric) | 15 cm |